Polysorbate 80 NF, Multi-Compendial, J.T.Baker®
Catalog # CAJT4117-3
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
CAS Number:
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Pack type:Glass bottle, amber, narrow mouth
- Size:4 L
- Cat. no.:CAJT4117-3
- Supplier No.:4117-03
Specifications
Specification Test Results
- Microbiological - Pseudomonas aeruginosa:Passes Test
- Microbiological - Salmonella:Passes Test
- Microbiological - Staphylococcus aureus:Passes Test
- Microbiological - Total Yeast and Mold Count (cfu/g):≤ 50
- Ethylene glycol:≤ 0.10 %
- Diethylene glycol :≤ 0.10 %
- NF - Acid Value:≤ 2.0
- NF - Hydroxyl Value:65 - 80
- NF - Identification A:Passes Test
- NF - Identification B:Passes Test
- NF - Residue on Ignition:≤ 0.25 %
- NF - Saponification Value:45 - 55
- NF - Specific Gravity at 25°C:1.06 - 1.09
- NF - Viscosity at 25°C, cSt:300 - 500
- NF - Water (H₂O):≤ 3.0 %
- NF - Peroxide Value:≤ 10.0
- NF - Ethylene Oxide:≤ 1 ppm
- NF - Dioxane:≤ 10 ppm
- NF - Composition of Fatty Acids - Myristic Acid:≤ 5.0 %
- NF - Composition of Fatty Acids - Palmitic Acid:≤ 16.0 %
- NF - Composition of Fatty Acids - Palmitoleic Acid:≤ 8.0 %
- NF - Composition of Fatty Acids - Stearic Acid:≤ 6.0 %
- NF - Composition of Fatty Acids - Oleic Acid:≥ 58.0 %
- NF - Composition of Fatty Acids - Linoleic Acid:≤ 18.0 %
- NF - Composition of Fatty Acids - Linolenic Acid:≤ 4.0 %
- EP - Acid Value:≤ 2.0
- EP - Total Ash:≤ 0.25 %
- EP - Hydroxyl Value:65 - 80
- EP - Identification A:Passes Test
- EP - Identification D:Passes Test
- EP - Peroxide Value:≤ 10.0
- EP - Ethylene Oxide:≤ 1 ppm
- EP - Dioxan:≤ 10 ppm
- EP - Saponification Value:45 - 55
- EP - Water (H₂O):≤ 3.0 %
- EP/BP - Composition of Fatty Acids - Myristic Acid:≤ 5.0 %
- EP/BP - Composition of Fatty Acids - Palmitic Acid:≤ 16.0 %
- EP/BP - Composition of Fatty Acids - Palmitoleic Acid:≤ 8.0 %
- EP/BP - Composition of Fatty Acids - Stearic Acid:≤ 6.0 %
- EP/BP - Composition of Fatty Acids - Oleic Acid:≥ 58.0 %
- EP/BP - Composition of Fatty Acids - Linoleic Acid:≤ 18.0 %
- EP/BP - Composition of Fatty Acids - Linolenic Acid:≤ 4.0 %
- Appearance:Passes Test
- JP - Acid Value:≤ 2.0
- JP - Composition of Fatty Acids - Myristic Acid:≤ 5.0 %
- JP - Composition of Fatty Acids - Palmitic Acid:≤ 16.0 %
- JP - Composition of Fatty Acids - Palmitoleic Acid:≤ 8.0 %
- JP - Composition of Fatty Acids - Stearic Acid:≤ 6.0 %
- JP - Composition of Fatty Acids - Oleic Acid:≥ 58.0 %
- JP - Composition of Fatty Acids - Linoleic Acid:≤ 18.0 %
- JP - Composition of Fatty Acids - Linolenic Acid:≤ 4.0 %
- JP - Dioxane:≤ 10 ppm
- JP - Ethylene Oxide:≤ 1 ppm
- JP - Heavy Metals (as Pb):≤ 20 ppm
- JP - Hydroxyl Value, meq KOH/g:65 - 80
- JP - Identification:Passes Test
- JP - Peroxide Value:≤ 10.0
- JP - Residue on Ignition:≤ 0.25 %
- JP - Water (H₂O):≤ 3.0 %
- JP - Saponification Value:45 - 55
- Additional Tests - Color (Gardner):≤ 7
- Additional Tests - Odor (Faint):Passes Test
- Additional Tests - Peroxide Value, meqO₂/kg:≤ 2.0
- Additional Tests - Endotoxin Concentration (EU/mL):≤ 10
- Free Ethylene Oxide:≤ 1 ppm
- 1,4-Dioxane:≤ 5 ppm
- Microbiological - Total Aerobic Microbial Count (cfu/g):≤ 100
- Microbiological - Escherichia Coli:Passes Test
Specification Test Results
About this item
:
Melting Pt:–21 °C
Density:1.062 g/cm³
:Management of Change (MOC) category = R
Flash Pt:113 °C (closed cup)
Storage Temperature:Ambient
EINECS:500-019-9
Frequently Bought Together