Potassium hydroxide, pellets NF, Multi-Compendial
Product Details & Documents
- CAS Number:1310-58-3
- Formula:KOH
- MDL Number:MFCD00003553
- Molecular Weight:56.11 g/mol
- Merck Index:12,07806
- Boiling Pt:1320 °C (1013 hPa)
- Melting Pt:360 °C
- Density:2.04 g/cm³ (20 °C)
- EINECS:215-181-3
- ADR:8,II
- :Management of Change (MOC) category = R
- Storage Temperature:Ambient
- UN:1813
- REACH:01-2119487136-33
Documents
Certificates
Search by Lot Number3152 Product Specifications VWR GHS Compliance Letter (US)
Additional Product Details
Avantor Portfolio of Chemicals
Product Literature
spec_JT3152-5
Product specification
Specifications
Specification Test Results
- NF - Identification A:Passes Test
- NF - Identification B:Passes Test
- NF - Insoluble Substances:Passes Test
- NF - Content of Potassium:≥59.9 %
- NF - Limit of Sodium:≤1.0 %
- NF - Assay (Total Alkali):≥85.0 %
- NF - Carbonate (as K₂CO₃):≤3.5 %
- EP/BP - Identification A:Passes Test
- EP/BP - Identification B:Passes Test
- EP/BP - Appearance of Solution:Passes Test
- EP/BP - Carbonate (as K₂CO₃):≤2.0 %
- EP/BP - Chloride (Cl):≤200 ppm
- EP/BP - Phosphate (PO₄):≤100 ppm
- EP/BP - Sulfate (SO₄):≤200 ppm
- EP/BP - Iron (Fe):≤10 ppm
- EP/BP - Sodium (Na):≤1.0 %
- EP/BP - Assay:85.0 - 100.5 %
- JP - Identification A:Passes Test
- JP - Identification B:Passes Test
- JP - Clarity and Color of Solution:Passes Test
- JP - Chloride (Cl):≤0.050 %
- JP - Heavy Metals (as Pb):≤30 ppm
- JP - Sodium (Na):Passes Test
- JP - Potassium Carbonate (K₂CO₃):≤2.0 %
- JP - Assay (KOH):≥85.0 %
Specifications
- Pack type:Poly bottle, white, blue T/E cap
- Size:500 g
Specifications
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



