L(+)-Ascorbic acid, GR ACS, Supelco®
100 g
Supplier: MilliporeSigma
Supplier #: AX1775-3
L(+)-Ascorbic acid, GR ACS, Supelco®
Catalog # CAAX1775-3
Supplier: MilliporeSigma
CAS Number: 50-81-7
Jump to:
Product Details & Documents
Synonyms: Antiscorbic vitamin, Ascorbic Acid, L-Ascorbic acid, Vitamin C, (R)-5-((S)-1,2-Dihydroxyethyl)-3,4-dihydroxyfuran-2(5H)-one, Acidum ascorbicum
- CAS Number:50-81-7
- Formula:C₆H₈O₆
- MDL Number:MFCD00064328
- Molecular Weight:176.13 g/mol
- Merck Index:13,00837
- Boiling Pt:553 °C (1013 hPa)
- Melting Pt:190…192 °C
- Density:1.694 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:200-066-2
- REACH:exempt
Documents
Specifications
Specification Test Results
- Assay:99.0% min
- Heavy metals (as Pb):0.002% max
- Iron (Fe):0.001ppm max
- Residue after ignition:0.1% max
- Specific rotation [a]25/D (10%, water):+20.5-+21.5°
Specifications
- Size:100 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



