Order Entry
Canada
Orders LinkContactUsLinkComponent
L(+)-Ascorbic acid, GR ACS, Supelco®

L(+)-Ascorbic acid, GR ACS, Supelco®

Plastic bottle, 250 g
Catalog # CAAX1775-2
L(+)-Ascorbic acid, GR ACS, Supelco®
Catalog # CAAX1775-2
Supplier:  
CAS Number:  50-81-7
L(+)-Ascorbic acid, GR ACS, Supelco®
L(+)-Ascorbic acid, GR ACS, Supelco®

Plastic bottle, 250 g

CAS Number:  50-81-7
Discontinued
Product Details & Documents
Synonyms:  Antiscorbic vitamin, Ascorbic Acid, L-Ascorbic acid, Vitamin C, (R)-5-((S)-1,2-Dihydroxyethyl)-3,4-dihydroxyfuran-2(5H)-one, Acidum ascorbicum
  • CAS Number:50-81-7
  • Formula:C₆H₈O₆
  • MDL Number:MFCD00064328
  • Molecular Weight:176.13 g/mol
  • Merck Index:13,00837
  • Boiling Pt:553 °C (1013 hPa)
  • Melting Pt:190…192 °C
  • Density:1.694 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:200-066-2
  • REACH:exempt
Specifications

Specification Test Results

  • Assay:
    99.0% min
  • Heavy metals (as Pb):
    0.002% max
  • Iron (Fe):
    0.001ppm max
  • Residue after ignition:
    0.1% max
  • Specific rotation [a]25/D (10%, water):
    +20.5-+21.5°

Specifications

  • Pack type:
    Plastic bottle
  • Size:
    250 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.