Celite® standard super-cel, filter aid
2.5 kg
Supplier: Thermo Scientific Chemicals
Supplier #: B21983-0E
Celite® standard super-cel, filter aid
Catalog # CAAAB21983-0E
Supplier: Thermo Scientific Chemicals
CAS Number: 91053-39-3
Jump to:
Product Details & Documents
CAS No.: 61790-53-2 Odor: Odorless Merck Reference: 14,4973 MDL No.: MFCD00132803
- CAS Number:91053-39-3
- MDL Number:MFCD00132803
- Merck Index:14,4973
- Boiling Pt:2200 °C (1013 hPa)
- Melting Pt:1710 °C
- Density:2.3 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:319-127-6
Documents
Specifications
Specifications
- Size:2.5 kg
Specifications
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.



