Order Entry
Canada
Orders LinkContactUsLinkComponent
Crystal violet

Crystal violet

Bottle, 500 g
Catalog # CAAAB21932-36
Corrosive
Irritant
Health
Environment
Supplier #:  B21932-36
Crystal violet
Catalog # CAAAB21932-36
CAS Number:  548-62-9
Crystal violet
Crystal violet

Bottle, 500 g

CAS Number:  548-62-9
Price
426,57 $
Log in for availability
Each (500g)
Minimum order quantity is 1
Product Details & Documents

CAS No.: 548-62-9 Molecular Formula: C25H30ClN3 Formula Weight: 407.99 Melting Point: ca 215° dec. Beilstein Reference: 4077708 Merck Reference: 14,4395 MDL No.: MFCD00011750

Synonyms:  Methylrosanilide chloride, Basic violet 3, Viocid, N-(4-(Bis(4-(dimethylamino)phenyl)methylene)cyclohexa-2,5-dien-1-ylidene)-N-methylmethanamonium chloride, Violet crystal, Methyl violet 10B, Dimethyl((di(4-(dimethylamino)phenyl)methylen)-2,5-cyclohexadien-1-ylidene)ammonium chloride, CI 42555, 4-[4,4'-Bis(dimethylamino) benzhydrylidene]cyclohexa-2,5-dien-1-ylidene]dimethylammonium chloride, Hexamethyl violet, Hexamethyl-p-rosaniline chloride, Vianin, Gentian violet
  • CAS Number:548-62-9
  • Formula:C₂₅H₃₀ClN₃
  • MDL Number:MFCD00011750
  • Molecular Weight:407.99 g/mol
  • Merck Index:14,4395
  • Melting Pt:∼ 215 °C (dec.)
  • Melting Pt:ca 215 °C (dec.)
  • Density:1.19 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:208-953-6
  • UN:3077
  • ADR:9,III
  • ADR:"9",III

Documents

Specifications

Specifications

  • Pack type:
    Bottle
  • Size:
    500 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.