Crystal violet
Bottle, 100 g
Supplier: Thermo Scientific Chemicals
Supplier #: B21932-22
Jump to:
Product Details & Documents
CAS No.: 548-62-9 Molecular Formula: C25H30ClN3 Formula Weight: 407.99 Melting Point: ca 215° dec. Beilstein Reference: 4077708 Merck Reference: 14,4395 MDL No.: MFCD00011750
Synonyms: Methylrosanilide chloride, Basic violet 3, Viocid, N-(4-(Bis(4-(dimethylamino)phenyl)methylene)cyclohexa-2,5-dien-1-ylidene)-N-methylmethanamonium chloride, Violet crystal, Methyl violet 10B, Dimethyl((di(4-(dimethylamino)phenyl)methylen)-2,5-cyclohexadien-1-ylidene)ammonium chloride, CI 42555, 4-[4,4'-Bis(dimethylamino) benzhydrylidene]cyclohexa-2,5-dien-1-ylidene]dimethylammonium chloride, Hexamethyl violet, Hexamethyl-p-rosaniline chloride, Vianin, Gentian violet
- CAS Number:548-62-9
- Formula:C₂₅H₃₀ClN₃
- MDL Number:MFCD00011750
- Molecular Weight:407.99 g/mol
- Merck Index:14,4395
- Melting Pt:∼ 215 °C (dec.)
- Melting Pt:ca 215 °C (dec.)
- Density:1.19 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:208-953-6
- UN:3077
- ADR:9,III
- ADR:"9",III
Documents
Specifications
Specifications
- Pack type:Bottle
- Size:100 g



