Order Entry
Canada
Orders LinkContactUsLinkComponent
ADA 98+%

ADA 98+%

Bottle, 25 g
Catalog # CAAAA15267-14
Supplier #:  A15267-14
ADA 98+%
Catalog # CAAAA15267-14
CAS Number:  26239-55-4
ADA 98+%

Bottle, 25 g

CAS Number:  26239-55-4
Price
$69.07
Log in for availability
Each (25g)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

Purity: 98+% CAS No.: 26239-55-4 Molecular Formula: H2NCOCH2N(CH2CO2H)2 Formula Weight: 190.16 Melting Point: ca 218° dec. Beilstein Reference: 1787181 Merck Reference: 14,147 MDL No.: MFCD00008031

Synonyms:  2,2'-[(2-Amino-2-oxoethyl)imino]diacetic acid, N-(2-Acetamido)iminodiacetic acid, 2,2'-((2-Amino-2-oxoethyl)azanediyl)diacetic acid
  • CAS Number:26239-55-4
  • Formula:H₂NC(O)CH₂N(CH₂COOH)₂
  • MDL Number:MFCD00008031
  • Molecular Weight:190.16 g/mol
  • Merck Index:14,147
  • Melting Pt:∼ 218 °C (dec.)
  • Storage Temperature:Ambient
  • EINECS:247-530-0

Documents

Specifications

Specifications

  • Pack type:
    Bottle
  • Size:
    25 g

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.