Order Entry
Canada
Orders LinkContactUsLinkComponent
 

trans-4-Hydroxycinnamic acid 98%

Bottle, 100 g
Catalog # CAAAA15167-22
Irritant
 :  A15167-22
trans-4-Hydroxycinnamic acid 98%
Catalog # CAAAA15167-22
CAS Number:  501-98-4
trans-4-Hydroxycinnamic acid 98%
trans-4-Hydroxycinnamic acid 98%

Bottle, 100 g

CAS Number:  501-98-4
Price
$421.82
Log in for availability
Each (100g)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

Purity: 98% CAS No.: 501-98-4 Molecular Formula: C9H8O3 Formula Weight: 164.16 Melting Point: ca 214° dec. Beilstein Reference: 2207383 Merck Reference: 14,2560 MDL No.: MFCD00004399

Synonyms:  p-Coumaric acid, trans-p-Cumaric Acid, trans-p-Coumaric Acid
  • CAS Number:501-98-4
  • Formula:C₉H₈O₃
  • MDL Number:MFCD00004399
  • Molecular Weight:164.16 g/mol
  • Merck Index:14,2560
  • Melting Pt:∼ 214 °C (dec.)
  • Storage Temperature:Ambient
  • EINECS:605-511-6
Specifications

Specifications

  • Pack type:
    Bottle
  • Size:
    100 g

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.