Order Entry
Canada
Orders LinkContactUsLinkComponent
1,6-Dibromohexane 97+%

1,6-Dibromohexane 97+%

Bottle, 100 g
Catalog # CAAAA13417-22
Irritant
Environment
Supplier #:  A13417-22
1,6-Dibromohexane 97+%
Catalog # CAAAA13417-22
CAS Number:  629-03-8
1,6-Dibromohexane 97+%

Bottle, 100 g

CAS Number:  629-03-8
Price
$53.60
Log in for availability
Each (100g)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

Purity: 97+% CAS No.: 629-03-8 Molecular Formula: Br(CH2)6Br Formula Weight: 243.98 Flash Point: 110°(230°F) Refractive Index: 1.5075 Odor: Odorless Melting Point: -2° Boiling Point: 242-244° Beilstein Reference: 1236322 UN No.: 3082 MDL No.: MFCD00000272

Synonyms:  Hexamethylen dibromide
  • CAS Number:629-03-8
  • Formula:BrH₂C(CH₂)₄CH₂Br
  • MDL Number:MFCD00000272
  • Molecular Weight:243.97 g/mol
  • Boiling Pt:242…244 °C
  • Melting Pt:–2 °C
  • Density:1.610
  • Flash Pt:>110 °C (230 °F)
  • Storage Temperature:Ambient
  • EINECS:211-067-2
  • UN:3082
  • ADR:9,III
Specifications

Specifications

  • Pack type:
    Bottle
  • Size:
    100 g

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.