Order Entry
Canada
Orders LinkContactUsLinkComponent
Formic acid 97%

Formic acid 97%

Bottle, 500 g
Catalog # CAAAA13285-36
Flammable
Corrosive
Toxic
Supplier #:  A13285-36
Formic acid 97%
Catalog # CAAAA13285-36
CAS Number:  64-18-6
Formic acid 97%
Formic acid 97%

Bottle, 500 g

CAS Number:  64-18-6
Price
80,60 $
Log in for availability
Each (500g)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

Purity: 97% CAS No.: 64-18-6 Molecular Formula: CH2O2 Formula Weight: 46.03 Flash Point: 50°(156°F) Refractive Index: 1.3710 Melting Point: 7-9° Boiling Point: 100-101° Beilstein Reference: 1209246 Merck Reference: 14,4241 UN No.: 1779 MDL No.: MFCD00003297

Synonyms:  Aminic Acid, Methanoic acid
  • CAS Number:64-18-6
  • Formula:HCO₂H
  • MDL Number:MFCD00003297
  • Molecular Weight:46.03
  • Merck Index:14,4241
  • Boiling Pt:100…101 °C
  • Melting Pt:7…9 °C
  • Density:1.220
  • Flash Pt:50 °C (156 °F)
  • Storage Temperature:Ambient
  • UN:1779
  • ADR:8,II
  •  :Density: 1.22 g/cm³ (25 °C)
  • EINECS:200-579-1

Documents

Specifications

Specifications

  • Pack type:
    Bottle
  • Size:
    500 g

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.