Order Entry
Canada
Orders LinkContactUsLinkComponent
1,6-Hexanediol 97%

1,6-Hexanediol 97%

Bottle, 500 g
Catalog # CAAAA12439-36
Supplier #:  A12439-36
1,6-Hexanediol 97%
Catalog # CAAAA12439-36
CAS Number:  629-11-8
1,6-Hexanediol 97%

Bottle, 500 g

CAS Number:  629-11-8
Price
$81.03
Log in for availability
Each (500g)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

Purity: 97% CAS No.: 629-11-8 Molecular Formula: HO(CH2)6OH Formula Weight: 118.18 Flash Point: 147°(296°F) Physical Form: Cryst solid Density: 1.120 Sensitivity: Hygroscopic Appearance: White Melting Point: 40-43° Boiling Point: 248-250° Beilstein Reference: 1633461 Merck Reference: 14,4690 MDL No.: MFCD00002985

Synonyms:  Hexamethylene glycol, 1,6-Dihydroxy hexane
  • CAS Number:629-11-8
  • Formula:C₆H₁₄O₂
  • MDL Number:MFCD00002985
  • Molecular Weight:118.18 g/mol
  • Merck Index:14,4690
  • Boiling Pt:248…250 °C
  • Melting Pt:40…43 °C
  • Density:1.120
  • Flash Pt:147 °C(296 °F)
  • Storage Temperature:Ambient
  • EINECS:211-074-0
Specifications

Specifications

  • Pack type:
    Bottle
  • Size:
    500 g

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.