Hydroxylammonium sulfate ≥99%
Product Details & Documents
MDL: MFCD00044869 Soluble in water
Purity: 99% CAS No.: 10039-54-0 Molecular Formula: H3NO.0.5H2SO4 Formula Weight: 164.14 Physical Form: Crystalline Density: 1.88 Appearance: White Odor: Odorless pH: 3.6 Melting Point: ca 170° dec. Solubility: Soluble in water Merck Reference: 14,4828 UN No.: 2865 MDL No.: MFCD00044869
- CAS Number:10039-54-0
- Formula:H₈N₂O₆S
- MDL Number:MFCD00044869
- Molecular Weight:164.14 g/mol
- Merck Index:14,4828
- Melting Pt:∼ 170 °C (dec.)
- Density:1.88
- Storage Temperature:Ambient
- EINECS:233-118-8
- UN:2865
- ADR:8,III
Specifications
Specifications
- Pack type:Bottle
- Size:2 kg
Specifications
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.



