Ninhydrin, REacton® ACS
Bottle, 25 g
Supplier: Thermo Scientific Chemicals
Supplier #: 43846-14
Ninhydrin, REacton® ACS
Catalog # CAAA43846-14
Supplier: Thermo Scientific Chemicals
CAS Number: 485-47-2
Jump to:
Product Details & Documents
Reagent for determination of amino acids and amine containing compounds, as an intermediate.
CAS No.: 485-47-2 Molecular Formula: C9H6O4 Formula Weight: 178.14 Physical Form: Crystalline Sensitivity: Light Sensitive Appearance: Pale yellow to pale brown Melting Point: 251° dec. Solubility: Soluble in water and alcohol Beilstein Reference: 1910963 Merck Reference: 14,6554 MDL No.: MFCD00003791
Reagent for determination of amino acids and amine containing compounds, as an intermediate.
Synonyms: Ninhydrin monohydrate, Trioxohydrindene monohydrate, 2,2-Dihydroxyindane-1,3-dione, 2,2-Dihydroxy-1,3-indanedione, Indanetrione hydrate, 2,2-Dihydroxyindan-1,3-dion, 1,2,3-Indantrione monohydrate
- CAS Number:485-47-2
- Formula:C₉H₆O₄
- MDL Number:MFCD00003791
- Molecular Weight:178.14 g/mol
- Merck Index:14,6554
- Boiling Pt:351 °C (1013 hPa)
- Melting Pt:251 °C (dec.)
- Density:0.862 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:207-618-1
Documents
Certificates
Search by Lot NumberSpecifications
Specification Test Results
- Appearance:White to brownish-white crystals
- Identification and melting point:Pass test.
- Solubility:Pass test
- Sensitivity to amino acids:Pass test
Specifications
- Pack type:Bottle
- Size:25 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



