Order Entry
Canada
Orders LinkContactUsLinkComponent
Buffer Solution, pH 7.00 (±0.01 @ 25 °C), No Color, Specpure®, NIST Traceable, Thermo Scientific

Buffer Solution, pH 7.00 (±0.01 @ 25 °C), No Color, Specpure®, NIST Traceable, Thermo Scientific

±0.01, 500 mL, 1.68, Colorless
Catalog # CAAA42415-AP
Supplier #:  42415-AP
Buffer Solution, pH 7.00 (±0.01 @ 25 °C), No Color, Specpure®, NIST Traceable, Thermo Scientific
Catalog # CAAA42415-AP
CAS Number:  
Buffer Solution, pH 7.00 (±0.01 @ 25 °C), No Color, Specpure®, NIST Traceable, Thermo Scientific

±0.01, 500 mL, 1.68, Colorless

CAS Number:  
Price
$59.91
Log in for availability
Each (500ml)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

NIST Traceable.

Physical Form: Liquid
Appearance: Colorless
MDL No.: MFCD03458582
Calibration of pH meters

Specifications

Specifications

  • Color:
    Colorless
  • Size:
    500 mL
  • pH:
    1.68
  • pH accuracy:
    ±0.01

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.