Order Entry
Canada
Orders LinkContactUsLinkComponent
 

Hafnium dioxide, tech.

Bottle, 100 g
Catalog # CAAA40270-22
 :  40270-22
Hafnium dioxide, tech.
Catalog # CAAA40270-22
CAS Number:  12055-23-1
Hafnium dioxide, tech.

Bottle, 100 g

CAS Number:  12055-23-1
Price
$412.91
Log in for availability
Each (100g)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

MDL: MFCD00003565

CAS No.: 12055-23-1 Molecular Formula: HfO2 Formula Weight: 210.49 Physical Form: -325 Mesh Powder Density: 9.68 Odor: Odorless Merck Reference: 14,4588 MDL No.: MFCD00003565

  • CAS Number:12055-23-1
  • Formula:HfO₂
  • MDL Number:MFCD00003565
  • Molecular Weight:210.49
  • Merck Index:14,4588
  • Boiling Pt:5400 °C (1013 hPa)
  • Melting Pt:2774 °C
  • Density:9.68
  •  :Density: 9.68 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:235-013-2
Specifications

Specifications

  • Pack type:
    Bottle
  • Size:
    100 g

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommandations

Les recommandations seront personnalisées selon vos préférences d’achat uniquement si vous avez activé "Améliorer mon expérience d’achat" dans la page "Informations personnelles".

Sinon, vous recevrez des recommandations génériques.