Order Entry
Canada
Orders LinkContactUsLinkComponent
 

D-(-)-Sorbitol 98%

Bottle, 500 g
Catalog # CAAA36404-36
 :  36404-36
D-(-)-Sorbitol 98%
Catalog # CAAA36404-36
CAS Number:  50-70-4
D-(-)-Sorbitol 98%
D-(-)-Sorbitol 98%

Bottle, 500 g

CAS Number:  50-70-4
Price
$70.11
Log in for availability
Each (500g)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

MDL: MFCD00004708 Soluble in water, methanol, acetone, DMF

Purity: 98% CAS No.: 50-70-4 Molecular Formula: C6H14O6 Formula Weight: 182.17 Physical Form: Powder Sensitivity: Hygroscopic Appearance: White Odor: Odorless Melting Point: 98-100° Solubility: Soluble in water, methanol, acetone, DMF Beilstein Reference: 1721899 Merck Reference: 14,8725 MDL No.: MFCD00004708

Synonyms:  (2R,3R,4R,5S)-Hexane-1,2,3,4,5,6-hexaol, (Sorbitol), D(-)-Glucitol, D-Sorbitol, Sorbitol
  • CAS Number:50-70-4
  • Formula:HOH₂C(CH(OH))₄CH₂OH
  • MDL Number:MFCD00004708
  • Molecular Weight:182.17 g/mol
  • Merck Index:14,8725
  • Boiling Pt:295 °C (1013 hPa)
  • Melting Pt:98…100 °C
  • Density:1.524 g/cm³ (20 °C)
  • Flash Pt:>100 °C
  • Storage Temperature:Ambient
  • EINECS:200-061-5
Specifications

Specifications

  • Pack type:
    Bottle
  • Size:
    500 g

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.