Order Entry
Canada
Orders LinkContactUsLinkComponent
L(+)-Ascorbic acid 99+% ACS

L(+)-Ascorbic acid 99+% ACS

Bottle, 25 g
Catalog # CAAA36237-14
Supplier #:  36237-14
L(+)-Ascorbic acid 99+% ACS
Catalog # CAAA36237-14
CAS Number:  50-81-7
L(+)-Ascorbic acid 99+% ACS
L(+)-Ascorbic acid 99+% ACS

Bottle, 25 g

CAS Number:  50-81-7
Price
$50.72
Log in for availability
Each (25g)
Minimum order quantity is 1
Product Details & Documents

MDL: MFCD00064328 Beilstein Registry No.: 84272

Purity: 99+% CAS No.: 50-81-7 Molecular Formula: C6H8O6 Formula Weight: 176.12 Physical Form: Powder Density: 1.65 Melting Point: ca 191° dec. Beilstein Reference: 84272 Merck Reference: 14,830 MDL No.: MFCD00064328

Synonyms:  Antiscorbic vitamin, Ascorbic Acid, L-Ascorbic acid, Vitamin C, (R)-5-((S)-1,2-Dihydroxyethyl)-3,4-dihydroxyfuran-2(5H)-one, Acidum ascorbicum
  • CAS Number:50-81-7
  • Formula:C₆H₈O₆
  • MDL Number:MFCD00064328
  • Molecular Weight:176.13 g/mol
  • Merck Index:14,830
  • Boiling Pt:553 °C (1013 hPa)
  • Melting Pt:∼ 191 °C (dec.)
  • Density:1.65
  • Storage Temperature:Ambient
  • EINECS:200-066-2
  • REACH:exempt

Documents

Specifications

Specification Test Results

  • Specific rotation ¬ +:
    21.0 +/-0.5°
  • Residue after ignition:
    0.1%
  • Heavy Metals (as Pb):
    0.002%
  • Fe:
    0.00001

Specifications

  • Pack type:
    Bottle
  • Size:
    25 g

Specifications