Order Entry
Canada
Orders LinkContactUsLinkComponent
 

Calcium fluoride 97%, REacton®, Reagent Grade

Bottle, 500 g
Catalog # CAAA33297-36
Irritant
 :  33297-36
Calcium fluoride 97%, REacton®, Reagent Grade
Catalog # CAAA33297-36
CAS Number:  7789-75-5
Calcium fluoride 97%, REacton®, Reagent Grade

Bottle, 500 g

CAS Number:  7789-75-5
Price
$118.18
Log in for availability
Each (500g)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

Spectroscopy, electronics, laser, high temperature dry-film lubricants

Purity: 97% CAS No.: 7789-75-5 Molecular Formula: CaF2 Formula Weight: 78.08 Physical Form: Powder Density: 3.18 Refractive Index: 1.434 Sensitivity: Hygroscopic Odor: Odorless Melting Point: 1403° Boiling Point: ca 2500° Solubility: Insoluble in water. Soluble in ammonium salts. Soluble in conc. mineral acids with evolution of HF Merck Reference: 14,1667 MDL No.: MFCD00010907

Spectroscopy, electronics, laser, high temperature dry-film lubricants

  • CAS Number:7789-75-5
  • Formula:CaF₂
  • MDL Number:MFCD00010907
  • Molecular Weight:78.07 g/mol
  • Merck Index:14,1667
  • Boiling Pt:∼2500 °C
  • Melting Pt:1403 °C
  • Density:3.18
  • Storage Temperature:Ambient
  • EINECS:232-188-7
Specifications

Specifications

  • Pack type:
    Bottle
  • Size:
    500 g

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.