Order Entry
Canada
Orders LinkContactUsLinkComponent
Bismuth(III) carbonate basic ≥98.5%

Bismuth(III) carbonate basic ≥98.5%

Bottle, 500 g
Catalog # CAAA33216-36
Supplier #:  33216-36
Bismuth(III) carbonate basic ≥98.5%
Catalog # CAAA33216-36
CAS Number:  5892-10-4
Bismuth(III) carbonate basic ≥98.5%

Bottle, 500 g

CAS Number:  5892-10-4
Price
$447.65
Log in for availability
Each (500g)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

Opacifier in X-ray diagnosis, enamel fluxes, ceramic glazes

Purity: 98.5% min CAS No.: 5892-10-4 Molecular Formula: (BiO)2CO3 (approximate) Formula Weight: 509.97 Physical Form: Powder Density: 6.86 Solubility: Practically insoluble in water, alcohol. Soluble in nitric acid, hydrochloric acid, concentrated acetic acid Merck Reference: 14,1282 MDL No.: MFCD00010877

Opacifier in X-ray diagnosis, enamel fluxes, ceramic glazes

Synonyms:  Bismuth carbonate oxide, Dibismuth carbonate dioxide, Bismuth subcarbonate
  • CAS Number:5892-10-4
  • Formula:(BiO)₂CO₃
  • MDL Number:MFCD00010877
  • Molecular Weight:509.97 g/mol
  • Merck Index:14,1282
  • Density:6.86
  • Storage Temperature:Ambient
  • EINECS:227-567-9

Documents

Specifications

Specifications

  • Pack type:
    Bottle
  • Size:
    500 g

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.