Order Entry
Canada
Orders LinkContactUsLinkComponent
Selenium dioxide ≥99.4% (metals basis) | Avantor

Selenium dioxide ≥99.4% (metals basis)

Bottle, 500 g
  CAAA12358-36
 
 
 
Fournisseur #:  12358-36
Selenium dioxide ≥99.4% (metals basis)
  CAAA12358-36
 :  7446-08-4
Selenium dioxide ≥99.4% (metals basis)

Bottle, 500 g

 :  7446-08-4
 
$438.05
   
Each (500g)
 
   
 

As an oxidizing agent, in manufacturing of other selenium compounds

Purity: 99.4% CAS No.: 7446-08-4 Molecular Formula: O2Se Formula Weight: 110.96 Physical Form: Powder Density: 3.95 Appearance: White to pink Odor: Odorless Melting Point: 340-350° subl. Solubility: Partially soluble in water, methanol, ethanol, acetone. Soluble in concentrated H2SO4 Merck Reference: 14,8434 UN No.: 3283 MDL No.: MFCD00003562

As an oxidizing agent, in manufacturing of other selenium compounds

  •  :7446-08-4
  •  :SeO₂
  •  :MFCD00003562
  •  :110.96 g/mol
  •  :14,8434
  •  :340…350 °C (subl.)
  •  :3.95
  •  :Ambient
  •  :231-194-7
  •  :3283
  •  :6.1,II

Documents

 

 

  • Pack type:
    Bottle
  • Size:
    500 g

 

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommandations

Les recommandations seront personnalisées selon vos préférences d’achat uniquement si vous avez activé "Améliorer mon expérience d’achat" dans la page "Informations personnelles".

Sinon, vous recevrez des recommandations génériques.