L(+)-Ascorbic acid 98+%
MDL: MFCD00064328 Beilstein Registry No.: 84272
Purity: 98+% CAS No.: 50-81-7 Molecular Formula: C6H8O6 Formula Weight: 176.12 Physical Form: Powder Density: 1.65 Melting Point: ca 191° dec. Beilstein Reference: 84272 Merck Reference: 14,830 MDL No.: MFCD00064328
- :50-81-7
- :C₆H₈O₆
- :MFCD00064328
- :176.13 g/mol
- :14,830
- :553 °C (1013 hPa)
- :∼ 191 °C (dec.)
- :1.65
- :Ambient
- :200-066-2
- :exempt
- Pack type:Bottle
- Size:100 g
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.



