Order Entry
Canada
Orders LinkContactUsLinkComponent
Electrode Fill Solutions

Electrode Fill Solutions

FRISCOLYT-B, Bottle, 25 ml, —
Catalog # CA97001-710
Supplier:   Mettler Toledo
Supplier #:  51343185
Electrode Fill Solutions
Catalog # CA97001-710
Supplier:  Mettler Toledo
CAS Number:  
Electrode Fill Solutions
Electrode Fill Solutions

FRISCOLYT-B, Bottle, 25 ml, —

CAS Number:  
Price
$34.50
Log in for availability
Each (25ml)
Product Details & Documents

FRISCOLYT-B™ (84.4% glycerol, 11% KCl, and 4.6% H₂O) is for low temperature samples.

Delivery information: Supplied in 25 ml (0.9 oz.) bottles.

Documents

Specifications

Specifications

  • Pack type:
    Bottle
  • Description:
    FRISCOLYT-B
  • Ion:
  • Volume:
    25 ml

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.