Order Entry
Canada
Orders LinkContactUsLinkComponent
Difco™ m Enterococcus Agar, BD Diagnostics | Avantor

Difco™ m Enterococcus Agar, BD Diagnostics

m Enterococcus Agar, 500 g
Catalog # CA90003-932
Supplier:   BD
Fournisseur #:  274620
Difco™ m Enterococcus Agar, BD Diagnostics
Catalog # CA90003-932
Supplier:  BD
CAS Number:  
Difco™ m Enterococcus Agar, BD Diagnostics

m Enterococcus Agar, 500 g

CAS Number:  
Price
$370.00
Log in for availability
Each (500g)
Product Details & Documents

Selective isolation and enumeration of enterococci using the membrane filtration technique.

Documents

Specifications

Specifications

  • Description:
    m Enterococcus Agar
  • Size:
    500 g

Specifications