Order Entry
Canada
Orders LinkContactUsLinkComponent
R2A Agar, BD

R2A Agar, BD

R2A Agar, 500 g
Catalog # CA90000-994
Supplier:   BD
Supplier #:  218263
R2A Agar, BD
Catalog # CA90000-994
Supplier:  BD
CAS Number:  
R2A Agar, BD

R2A Agar, 500 g

CAS Number:  
Price
$355.00
Log in for availability
Each (500g)
Product Details & Documents

R2A Agar is used for enumerating heterotrophic organisms in treated potable water.

  • Dehydrated appearance: Light beige, flree-flowing
  • Prepared appearance: Light amber, slightly opalescent
  • Improves recovery of chlorine-tolerant bacteria

A low-nutrient medium, R2A Agar is used in combination with a lower incubation temperature and longer incubation time to stimulate the growth of stressed and chlorine-tolerant bacteria.

Specifications

Specifications

  • Description:
    R2A Agar
  • Size:
    500 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.