R2A Agar, BD
R2A Agar, 100 g
Jump to:
Discontinued
Product has been discontinued. However we found similar alternative products.
Alternatives
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
R2A Agar is used for enumerating heterotrophic organisms in treated potable water.
- Dehydrated appearance: Light beige, flree-flowing
- Prepared appearance: Light amber, slightly opalescent
- Improves recovery of chlorine-tolerant bacteria
A low-nutrient medium, R2A Agar is used in combination with a lower incubation temperature and longer incubation time to stimulate the growth of stressed and chlorine-tolerant bacteria.
Specifications
Specifications
- Description:R2A Agar
- Size:100 g