Order Entry
Canada
Orders LinkContactUsLinkComponent
Inoculum Broth, BD

Inoculum Broth, BD

Difco™ Micro Inoculum Broth, 500 g
Catalog # CA90000-366
Supplier:   BD
Supplier #:  211813
Inoculum Broth, BD
Catalog # CA90000-366
Supplier:  BD
CAS Number:  
Inoculum Broth, BD

Difco™ Micro Inoculum Broth, 500 g

CAS Number:  
Price
$662.00
Log in for availability
Each (500g)
Product Details & Documents

Inoculum Broth is used for preparing the inoculum of lactobacilli and other microorganisms used in microbiological assays of vitamins and amino acids.

  • Used for cultivating lactobacilli
  • For preparing the inoculum for microbiological assays
  • Peptone provides nitrogen and amino acids
  • Beige, homogenous appearance when dehydrated

Inoculum Broth is the recommended medium for preparation of lactobacilli inoculum for vitamin assays.

Specifications

Specifications

  • Description:
    Difco™ Micro Inoculum Broth
  • Size:
    500 g

Specifications