Tissue Capsules, Simport
28×5 mm
Supplier: Simport Scientific
Supplier #: M470
Jump to:
Product Details & Documents
These polypropylene capsules are suitable for holding tissue samples during processing
The lids have a frosted marking area for sample identification and an open mesh area to facilitate fluid exchange. The whole surface of the base is also an open mesh. The lid snaps securely on the base, eliminating the risk of tissue loss during processing. Color: white.
Documents
Certificates
Search by Lot NumberSpecifications
- Diameter ×H:28×5 mm