Order Entry
Canada
Orders LinkContactUsLinkComponent
StatSpin® Microhematocrit Tubes, Iris Sample Processing

StatSpin® Microhematocrit Tubes, Iris Sample Processing

SafeCrit plastic capillary tube, Sodium Heparin anticoagulant, 75 mm, 40 µl
Catalog # CA56430-048
Supplier:   Statspin
Supplier #:  HP4H-10
StatSpin® Microhematocrit Tubes, Iris Sample Processing
Catalog # CA56430-048
Supplier:  Statspin
StatSpin® Microhematocrit Tubes, Iris Sample Processing
StatSpin® Microhematocrit Tubes, Iris Sample Processing

SafeCrit plastic capillary tube, Sodium Heparin anticoagulant, 75 mm, 40 µl

Price
$42.04
Log in for availability
Pack of 100 (1/Itm/Bx) Case of 1000 (1/ Itm/Bx)
Product Details & Documents

These microhematocrit tubes use capillary action to draw samples quickly.

  • Capillary Action Draws Samples Quickly
  • Material: Non-Breakable Plastic for Safety
  • Heparin Tubes for Finger-Stick Samples
  • Untreated Tubes for Anti-Coagulated Venous Samples
  • Available Lengths: 75 mm & 40 mm

Tubes are available with heparin for finger-stick samples or untreated for anti-coagulated venous samples

SafeCrit® hematocrit tubes are made of non-breakable, 100% plastic to eliminate the potential safety hazards of broken glass.

The 40 mm tubes are made for use with the MP multipurpose centrifuge (87002-248 series) and the CritSpin™ microhematocrit centrifuge (87002-206 series). The 75 mm tubes are made for use in conventional centrifuges that support the full length of the tube.

Specifications
  • Volume:
    40 µl
  • Description:
    SafeCrit plastic capillary tube, Sodium Heparin anticoagulant
  • Length:
    75 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.