Order Entry
Canada
Orders LinkContactUsLinkComponent
StatSpin® Microhematocrit Tubes, Iris Sample Processing

StatSpin® Microhematocrit Tubes, Iris Sample Processing

SafeCrit plastic capillary tube, Sodium Heparin anticoagulant, 40 mm, 20 µl
Catalog # CA56430-046
Supplier:   Statspin
Supplier #:  HP8H-10
StatSpin® Microhematocrit Tubes, Iris Sample Processing
Catalog # CA56430-046
Supplier:  Statspin
StatSpin® Microhematocrit Tubes, Iris Sample Processing
StatSpin® Microhematocrit Tubes, Iris Sample Processing

SafeCrit plastic capillary tube, Sodium Heparin anticoagulant, 40 mm, 20 µl

Price
$58.32
Log in for availability
Pack of 100 Case of 1000
Product Details & Documents

These microhematocrit tubes use capillary action to draw samples quickly.

  • Capillary Action Draws Samples Quickly
  • Material: Non-Breakable Plastic for Safety
  • Heparin Tubes for Finger-Stick Samples
  • Untreated Tubes for Anti-Coagulated Venous Samples
  • Available Lengths: 75 mm & 40 mm

Tubes are available with heparin for finger-stick samples or untreated for anti-coagulated venous samples

SafeCrit® hematocrit tubes are made of non-breakable, 100% plastic to eliminate the potential safety hazards of broken glass.

The 40 mm tubes are made for use with the MP multipurpose centrifuge (87002-248 series) and the CritSpin™ microhematocrit centrifuge (87002-206 series). The 75 mm tubes are made for use in conventional centrifuges that support the full length of the tube.

Specifications
  • Volume:
    20 µl
  • Description:
    SafeCrit plastic capillary tube, Sodium Heparin anticoagulant
  • Length:
    40 mm