Order Entry
Canada
ContactUsLinkComponent
DPBS 1X WITHOUT CA+MG PK 6 500ML
Dulbecco's phosphate-buffered saline (DPBS), Mediatech
Catalog # CA45000-434
Supplier:  Corning
CAS Number:  
Dulbecco's phosphate-buffered saline (DPBS), Mediatech
Catalog # CA45000-434
Supplier:  Corning
Supplier Number:  21-031-CV
CAS Number:  

Specifications

  • Description:
    DPBS, 1X without calcium and magnesium
  • Storage temperature:
    Ambient
  • Cat. no.:
    CA45000-434
  • Storage Temperature:
    +2 °C…+30 °C
  • Volume:
    500 ml
  • Supplier No.:
    21-031-CV

Specifications

About this item

Dulbecco's Phosphate Buffered Saline (DPBS) is a buffer solution which can be used to maintain cell culture media in the physiological pH range of 7.0 to 7.6

  • Low endotoxin - < 0.25EU/mL
  • Mycoplasma tested
  • Liquid media products are formulated with cell culture grade WFI water
  • Manufactured in a cGMP environment

Complete Certificate of Analysis available for each production lot, along with full formulation information.

It can serve as an irrigating, transporting or diluting fluid while maintaining cell tonicity and viability for a limited period of time.

Caution: Storage temperature: Solutions 2 - 30 °C; Powder formats 2 - 8 °C
Frequently Bought Together