Orders LinkContactUsLinkComponent
Corning® FluoroBlok™ Multiwell Insert Systems, Corning
Corning® FluoroBlok™ Multiwell Insert Systems, Corning
24, 8.0 µm
Catalog # CA12222-502
Supplier:   Corning
Supplier #:  351157
Corning® FluoroBlok™ Multiwell Insert Systems, Corning
Catalog # CA12222-502
Supplier:  Corning
Corning® FluoroBlok™ Multiwell Insert Systems, Corning
Corning® FluoroBlok™ Multiwell Insert Systems, Corning
24, 8.0 µm
Price
$353.00
Log in for availability
Each
Minimum order quantity is 1
Specifications
  • No. of wells:
    24
  • Pore size:
    8.0 µm
Accessories
Product Information
  • Catalog #
    Availability
    Price
  • Square-Well, Flat-Bottom Assay Plates for 96-Well Systems
    Square-Well, Flat-Bottom Assay Plates for 96-Well Systems
    CABD353928
    Log in for availability
    Case of 5 - $276.00