Orders LinkContactUsLinkComponent
Replaceable Junction Tool and Ceramic Junctions, METTLER TOLEDO®
Replaceable Junction Tool and Ceramic Junctions, METTLER TOLEDO®
Replaceable Junction Tool
Catalog # CA11278-736
Supplier:   Mettler Toledo
Supplier #:  51340267
Replaceable Junction Tool and Ceramic Junctions, METTLER TOLEDO®
Catalog # CA11278-736
Supplier:  Mettler Toledo
Replaceable Junction Tool and Ceramic Junctions, METTLER TOLEDO®
Replaceable Junction Tool and Ceramic Junctions, METTLER TOLEDO®
Replaceable Junction Tool
Price
$232.00
Log in for availability
Each
Product Details & Documents
With a simple technique, this tool quickly replaces old, contaminated ceramic junctions from the InLab® 403 electrodes.
Specifications
  • Description:
    Replaceable Junction Tool