Order Entry
Canada
Orders LinkContactUsLinkComponent
qScript™ cDNA SuperMix, QuantaBio

qScript™ cDNA SuperMix, QuantaBio

qScript™ cDNA SuperMix, 5X, 100
Catalog # CA101414-104
Supplier:   Quantabio
Supplier #:  95048-100
qScript™ cDNA SuperMix, QuantaBio
Catalog # CA101414-104
Supplier:  Quantabio
CAS Number:  
qScript™ cDNA SuperMix, QuantaBio
qScript™ cDNA SuperMix, QuantaBio

qScript™ cDNA SuperMix, 5X, 100

CAS Number:  
Price
$906.77
Log in for availability
Each
Minimum order quantity is 1
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

SuperMix provides a sensitive and easy-to-use solution for two-step qRT-PCR

  • Sensitive first-strand cDNA synthesis of RNA sequences ≤1kb
  • Single tube, 5X concentrated master mix minimizes pipetting and improve assay accuracy
  • Broad linear dynamic detection range with limiting (10 pg) or plentiful (1 µg) samples of total RNA
  • Pre-blended with ribonuclease inhibitor protein (RIP) to prevent RNA template degradation during incubation and a proprietary blend of random and oligo(dT) primers, optimized to deliver unbiased representation of 5’ and 3’ sequences

This 5x concentrated master mix provides all necessary components (except RNA template) for first-strand synthesis including: buffer, dNTPs, MgCl2, primers, RNase inhibitor protein, qScript reverse transcriptase, and stabilizers. qScript™i s a RNase H(+) derivative of MMLV-RT, optimized for reliable cDNA synthesis over a wide dynamic range of input RNA. The unique blend of oligo (dT) and random primers in the qScript™ cDNA SuperMix works exceptionally well with a wide variety of targets. This blend is optimized for the production of targets <1kb in length. qScript™ cDNA SuperMix produces excellent results in both real-time and conventional RT-PCR.

Specifications

Specifications

  • Concentration:
    5X
  • Description:
    qScript™ cDNA SuperMix
  • No. of reactions:
    100

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.