Order Entry
Canada
Orders LinkContactUsLinkComponent
BBL™ Saline, Normal, BD

BBL™ Saline, Normal, BD

BBL™ saline, normal, 100×1 ml, Size K
Catalog # CA-90005-030
Supplier:   BD
Supplier #:  297815
BBL™ Saline, Normal, BD
Catalog # CA-90005-030
Supplier:  BD
CAS Number:  
BBL™ Saline, Normal, BD

BBL™ saline, normal, 100×1 ml, Size K

CAS Number:  
Price
300,00 $
Log in for availability
Pack of 100 (1ml)
Minimum order quantity is 1
Product Details & Documents

Normal saline is used in procedures that require the use of an isotonic diluent.

  • Ready to use
  • Provide ensured cell integrity
  • Used with a vast array of procedures

An isotonic diluent may be used for dilution of bacterial cells to provide a concentration suitable for microscopic observation, determination of cell numbers, analysis for genetic or metabolic properties, washing cells preparatory to study, or preparation of standardized inocula.

Specifications

Specifications

  • Description:
    BBL™ saline, normal
  • Size:
    100×1 ml
  • Tube size:
    Size K

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.