VWR® Glove Liners
Half Finger Liners, with Orange Cuff, Medium, White
Supplier: VWR International
Jump to:
Product Details & Documents
Ergonomically designed glove liners wick away moisture for comfortable use with disposable gloves.
- Ambidextrous glove liners
- Reduces perspiration and wicks away moisture
- Low-lint, 100% 15 denier nylon composition
- Sturdy, launderable, and reusable
- Color-coded cuffs indicate size
- Pre-laundered; will not shrink
Liners provide protection and comfort when used with latex, nitrile, and PVC gloves. Fully launderable for reusability and economy.
Packaging: Glove Liners are packaged 20 pair per bag, 15 bags per case. They are packed in clean polyethylene bags that are free of silicone, dioctyl phthalate (DOP), and amides.
Documents
Specifications
- Glove color:White
- Glove size:Medium
- Description:Half Finger Liners, with Orange Cuff
- Cat. No.:94000-618
- Disposable / reusable:Reusable
- Ambidextrous/Hand-specific:Ambidextrous
- Cuff style:Knit wrist