VWR® Glove Liners
Full finger liners, with blue cuff, Large, White
Ergonomically designed glove liners wick away moisture for comfortable use with disposable gloves.
- Ambidextrous glove liners
- Reduces perspiration and wicks away moisture
- Low-lint, 100% 15 denier nylon composition
- Sturdy, launderable, and reusable
- Color-coded cuffs indicate size
- Pre-laundered; will not shrink
Liners provide protection and comfort when used with latex, nitrile, and PVC gloves. Fully launderable for reusability and economy.
: Glove Liners are packaged 20 pair per bag, 15 bags per case. They are packed in clean polyethylene bags that are free of silicone, dioctyl phthalate (DOP), and amides.
- Glove color:White
- Glove size:Large
- Description:Full finger liners, with blue cuff
- Cat. No.:94000-612
- Disposable / reusable:Reusable
- Ambidextrous/Hand-specific:Ambidextrous
- Cuff style:Knit wrist