Pipette Tips, Sorenson BioScience
Extended length, 1 - 300 µl, Natural
Catalog # 89082-378
Supplier: Sorenson Bioscience
Supplier #: 30920
Jump to:
Product Details & Documents
Sorenson BioScience universal fit pipette tips are manufactured from medical grade polypropylene in a tightly controlled production environment.
Certified free of RNase / DNase, Human DNA, Pyrogens and PCR Inhibitors.
The accurate and precise tips are designed to fit most research-grade pipettors. Available in low binding, pipette tips reduce sample retention.
Specifications
- Volume:1 - 300 µl
- Tip color:Natural
- Tip style:Extended length