Orders LinkContactUsLinkComponent
Septa, Flat, Microscale, Ace Glass Incorporated
Septa, flat, for 5/5 thread joint, 8 mm
  89064-072
 :   Ace Glass
 :  8787-40
Septa, Flat, Microscale, Ace Glass Incorporated
  89064-072
 :  Ace Glass
Septa, Flat, Microscale, Ace Glass Incorporated
Septa, Flat, Microscale, Ace Glass Incorporated
Septa, flat, for 5/5 thread joint, 8 mm
$137.54
Case of 48
Replacement flat septa are designed for use with microscale reaction vials, or any other V-Bottom reaction vials.
These septa are constructed of white silicone rubber, bonded with a white PTFE facing. They will resist coring when punctured by a syringe needle. All septa are 3 mm thick.
  • Ø:
    8 mm
  • Description:
    Septa, flat, for 5/5 thread joint