Order Entry
Canada
Orders LinkContactUsLinkComponent
Axygen® Tip Refill System, Empty Racks, Corning

Axygen® Tip Refill System, Empty Racks, Corning

Empty racks; 10 racks/pack, 5 packs/case, 10, 200 µl Axygen® tips
Catalog # 89029-922
Supplier:   Corning
Supplier #:  RFL-R
Axygen® Tip Refill System, Empty Racks, Corning
Catalog # 89029-922
Supplier:  Corning
Axygen® Tip Refill System, Empty Racks, Corning

Empty racks; 10 racks/pack, 5 packs/case, 10, 200 µl Axygen® tips

Price
$35.60
Log in for availability
Pack of 10 Case of 50
Product Details & Documents

Axygen® Tip Refill System contains durable refill rack that can be easily refilled by hand and is autoclavable.

  • RFL-R-1200 is suitable for 1200 µl refill inserts
  • RFL-R is suitable for 10 and 200 µl refill inserts
  • RNase- and DNase-free, non pyrogenic
  • Translucent rack allows tips to be easily seen
  • Color-coded insert allows for easy identification of tip

They are available in different colors and are packaged in bulk or racks.

Documents

Specifications
  • Description:
    Empty racks; 10 racks/pack, 5 packs/case
  • For use with:
    10, 200 µl Axygen® tips
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.