$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Canada
ContactUsLinkComponent
GASKET 1.5 EP TAB ORI. EPDM
Catalog # 89011-136
Supplier:
Rubber Fab
Expand image
GASKET 1.5 EP TAB ORI. EPDM
Catalog # 89011-136
Supplier:
Rubber Fab
Supplier Number:
A80MPE-150-TAB-EP
Frequently Bought Together
pH Reference Standard Buffers, VWR Chemicals BDH®
Supplier: BDH
VWR® Disposable Petri Dishes
Supplier: VWR International
Falcon® Centrifuge Tubes, Conical Bottom, Corning
Supplier: Corning
VWR® Nitrile Examination Gloves, Blue
Supplier: VWR International
VWR® Light-Duty Tissue Wipers
Supplier: VWR International
VWR® Universal Pipette Tips
Supplier: VWR International
VWR® Microcentrifuge Tubes, PP
Supplier: VWR International