Orders LinkContactUsLinkComponent
Test Tubes without Rim
Test Tubes without Rim
10 ml, 15×125 mm
Catalog # 82024-106
Supplier #:  TT9820-D
Test Tubes without Rim
Catalog # 82024-106
Test Tubes without Rim
Test Tubes without Rim
10 ml, 15×125 mm
Price
$123.40
Log in for availability
Pack of 72 Case of 720
Product Details & Documents
Pre-cleaned borosilicate glass test tubes without rims. Tubes can be used for hot or cold applications, are reusable, and can be autoclaved.
  • Tubes do not have rims
  • Autoclavable
Rimless borosilicate glass test tubes are ideal for a variety of laboratory applications.
Specifications
  • Capacity:
    10 ml
  • Diameter ×H:
    15×125 mm