Orders LinkContactUsLinkComponent
Nunc® Barcode Scanners
Nunc® Barcode Scanners
Catalog # 73521-418
Supplier #:  330050
Nunc® Barcode Scanners
Catalog # 73521-418
Supplier:  
Nunc® Barcode Scanners
Nunc® Barcode Scanners
Discontinued
Product has been discontinued. However we found similar alternative products.
Alternatives
BARCODE SCANNER NUNC USB CS1 Catalog #CA73521-418
Product CA73521-418 is a replacement for 73521-418 and has the same packaging and specifications, but may be a different quantity.
Product Details & Documents
Nunc bar code scanners with plug and play functionality play a critical role in sample tracking
The scanners automatically switch between code 128, code 39, interleave 2 of 5 and several other barcodes. These scanners are durable low-cost, and easy to use.