Data-Trend Multipurpose Temperature/Humidity Chart Recorders, Dickson®
Product Details & Documents
Adaptable to a wide range of applications, these recorders feature a 15.2cm (6") readable chart, AC power with battery backup, and user-selectable recording times and temperature ranges
Increased shielding eliminates electronic noise interference and allows smoother pen movement.
Ordering information: Recorders include one red pen, one blue pen, AC adapter, and 9V battery. Charts must be purchased separately.
Services: VWRCATALYST™ offers validation services including IQ, OQ, and PQ protocol development and execution, as well as asset management and calibration services (US only); call 888-793-2300 for more details.
Specifications
- Temperature range:0...50 °C
- Description:7-Day Chart
Accessories
Product Information
- Catalog #AvailabilityPrice
- NIST Traceable Calibration 1-Point Service
Product Information
- Catalog #AvailabilityPrice
- NIST & A2LA Traceable Calibration 3-Point Service
Product Information
- Catalog #AvailabilityPrice
- Pens, Three Red and Three Blue
Les recommandations seront personnalisées selon vos préférences d’achat uniquement si vous avez activé "Améliorer mon expérience d’achat" dans la page "Informations personnelles".
Sinon, vous recevrez des recommandations génériques.