Laser Pointer, Sper Scientific
Catalog # 61220-788
Supplier: Sper Scientific
Specifications
- Description:Laser pointer
- Cat. no.:61220-788
- Supplier No.:330001
Specifications
About this item
Sturdy construction and a long battery life allow for heavy use as a teaching tool for science presentations, conferences, lectures, seminars, and planetariums
Designed with an attractive, brushed-metal finish, this incredibly thin pen-shaped unit can be easily carried in a shirt pocket. It projects a brilliant red dot clearly visible even in bright ambient light or with video, film, and slide presentations. Runs for a minimum of 4 continuous hours on 2 standard alkaline batteries (included).
Frequently Bought Together