Orders LinkContactUsLinkComponent
VWR® Caps
VWR® Caps
Red
Catalog # 60828-762
Supplier:   VWR International
VWR® Caps
Catalog # 60828-762
Supplier:  VWR International
VWR® Caps
VWR® Caps
Red
Price
$304.24
New Customer Promotional Price
Log in for availability
Pack of 1,000 Case of 10000
Product Details & Documents
Safe-T-Flex caps feature a unique, flexible, over-locking design that locks securely on the outside of the tube lip and can be removed with one hand.
  • Polyethylene construction
  • Liquid-tight seal
  • Color-coded for easy specimen identification
  • Cap top can be labeled
  • Self-locking caps hold firmly in any position
A tight seal ensures sample integrity and prevents spillage or evaporation of serum samples
Caps fit all varieties of blood collecting and disposable culture tubes. They are color-coded for easy specimen identification and to help guard against cross-contamination. Top of cap can also be numbered or labelled with indelible ink. Self-locking caps hold firmly in any position and keep contents secure while travelling. Liquid-tight seal withstands centrifugation, agitation, refrigeration, and freezing. Manufactured from polyethylene.
This special seal protects against the aerosols of highly infectious organisms such as TB and the HTLV-III virus.
Documents
Specifications
  • Color:
    Red