Order Entry
Canada
Orders LinkContactUsLinkComponent
VWR® Bacti Cell Spreaders

VWR® Bacti Cell Spreaders

Yellow, 24.0 cm, 6.0 cm, Non sterile
Catalog # 60828-684
Supplier:   VWR International
VWR® Bacti Cell Spreaders
Catalog # 60828-684
Supplier:  VWR International
VWR® Bacti Cell Spreaders
VWR® Bacti Cell Spreaders

Yellow, 24.0 cm, 6.0 cm, Non sterile

Price
$203.00
Log in for availability
Pack of 25 Case of 100
Product Details & Documents

Convenient and economical cell spreaders are designed to simplify bacterial culture work.

  • Autoclavable
  • Simplify bacterial culture work
  • Durable fabrication

Durable enough to be autoclaved and reused.

Ordering information: Cell spreaders are available sterile (individually wrapped) or nonsterile (bulk packed).

Documents

Certificates
Search by Lot Number
Specifications
  • Sterility:
    Non sterile
  • Color:
    Yellow
  • Blade width:
    6.0 cm
  • Overall length:
    24.0 cm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.