Order Entry
Canada
Orders LinkContactUsLinkComponent
VWR® Plug-Type Caps, PE

VWR® Plug-Type Caps, PE

Natural
Catalog # 60819-003
   Sustainable Packaging|Low Manufacturing Impact
Supplier:   VWR International
VWR® Plug-Type Caps, PE
Catalog # 60819-003
Supplier:  VWR International
   Sustainable Packaging|Low Manufacturing Impact
VWR® Plug-Type Caps, PE
VWR® Plug-Type Caps, PE

Natural

Price
$243.08
New Customer Promotional Price
Log in for availability
Pack of 1,000 Case of 3000
Product Details & Documents

Disposable caps are for 12 x 75 mm and 13 x 100 mm culture tubes.

  • Disposable
  • Polyethylene construction
  • Unique fluted design
  • Snugly grasps plastic or glass tubes

They feature a unique fluted design that grips plastic or glass tubes snugly.

Ordering information: The assorted pack contains 500 caps each in yellow, orange, red, blue, green and purple.

Specifications
  • Color:
    Natural
  • Cat. No.:
    60819-003
  • Environmentally Preferable:
    Sustainable Packaging|Low Manufacturing Impact
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.