Order Entry
Canada
Orders LinkContactUsLinkComponent
Stopcocks, plain bore, PYREX® Rotaflo®

Stopcocks, plain bore, PYREX® Rotaflo®

13×224 mm, 0 - 10 mm (0 - 0.394")
Catalog # 59285-182
Supplier:   Corning
Supplier #:  7470-10
Stopcocks, plain bore, PYREX® Rotaflo®
Catalog # 59285-182
Supplier:  Corning
Stopcocks, plain bore, PYREX® Rotaflo®
Stopcocks, plain bore, PYREX® Rotaflo®

13×224 mm, 0 - 10 mm (0 - 0.394")

Price
$381.00
Log in for availability
Case of 1
Product Details & Documents

PYREX®, borosilicate glass.

  • Fine flow adjustment
  • Lubrication is not required
  • Fast assembly

These high performance Rotaflo® stopcocks can be adjusted with bore openings and have sidearms. Only chemically resistant PTFE and PYREX® borosilicate glass are in contact with liquids and gase. Seal is visible when open or closed.

Documents

Specifications
  • Sidearm Dimensions (Ø×L):
    13×224 mm
  • Bore size:
    0 - 10 mm (0 - 0.394")
Recommandations

Les recommandations seront personnalisées selon vos préférences d’achat uniquement si vous avez activé "Améliorer mon expérience d’achat" dans la page "Informations personnelles".

Sinon, vous recevrez des recommandations génériques.