Orders LinkContactUsLinkComponent
Staticide® Antistatic Solution, ACL Staticide
Staticide® Antistatic Solution, ACL Staticide
General-Purpose, 3.8 L (1 gal.)
Catalog # 58611-327
Supplier:   ACL Staticide
Supplier #:  2001
Staticide® Antistatic Solution, ACL Staticide
Catalog # 58611-327
Supplier:  ACL Staticide
Staticide® Antistatic Solution, ACL Staticide
General-Purpose, 3.8 L (1 gal.)
Price
$168.77
Log in for availability
Each(1/Itm/Bx) Case of 4 (1/ Itm/Bx)
Product Details & Documents
Easy-to-use topical antistatic solution for long-term static control
Choose from pour-top gallons or ready-to-use quart bottles with sprayer. Cleanroom formula is individually bagged to prevent contamination.
Apply to floors, walls, ceilings, fixtures, work surfaces, carpets, and more. Effective in PC-board solder fault reduction programs; used in high sensitivity electronic and EDP operations. Nontoxic and nonflammable. Completely biodegradable
Certifications: Cleanroom formula is ISO Class 5 (FED-STD-209E Class 100/M3.5) approved. Meets MIL-B-81705B and NFPA-56A electrostatic decay criteria.
Documents
Specifications
  • Description:
    General-Purpose
  • Size:
    3.8 L (1 gal.)